Assay Design
Gene Transcripts
- Gene Symbol
- HOTAIRM1
- Entrez Gene ID
-
100506311
- Gene Name
- HOXA transcript antisense RNA, myeloid-specific 1
- Gene Aliases
-
HOXA-AS1, HOXA1-AS1, NCRNA00179
- Chromosome Location
-
Chr.7: 27096094 - 27100258 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_038366.1 |
|
2 - 3 |
563 |
66 |
GenBank mRNA |
BC031342.1 |
- |
2 - 3 |
533 |
66 |
View Full Product Details