Assay Design



Gene Transcripts

Gene Symbol
SNORA53
Entrez Gene ID
677832
Gene Name
small nucleolar RNA, H/ACA box 53
Gene Aliases
ACA53
Chromosome Location
Chr.12: 98599635 - 98599884 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_003015.1 1 - 1 149 136


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream thymopoietin TMPO CMD1T, LAP2, LEMD4, PRO0868, TP -49.3kb Sense Chr. - 98515573 - 98550379 on Build GRCh38
Downstream IKBKB interacting protein IKBIP IKIP 13.5kb Anti-sense Chr. - 98613404 - 98645051 on Build GRCh38

View Full Product Details