Assay Design



Gene Transcripts

Gene Symbol
CT62
Entrez Gene ID
196993
Gene Name
cancer/testis antigen 62
Gene Aliases
-
Chromosome Location
Chr.15: 71110244 - 71115551 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001102658.1 NP_001096128.1 3 - 4 645 81
XM_006720429.3 XP_006720492.1 3 - 4 905 81
GenBank mRNA BC039359.1 - 3 - 4 641 81
BC048128.1 - 3 - 4 607 81

View Full Product Details