Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001114099.2 | NP_001107571.1 | 7 - 8 | 1242 | 94 |
NM_001114100.2 | NP_001107572.1 | 5 - 6 | 1110 | 94 | |
NM_001243332.1 | NP_001230261.1 | 7 - 8 | 1452 | 94 | |
NM_001243333.1 | NP_001230262.1 | 6 - 7 | 1320 | 94 | |
NM_012410.3 | NP_036542.1 | 7 - 8 | 1242 | 94 | |
NM_201575.3 | NP_963869.2 | 7 - 8 | 1452 | 94 | |
XM_005255252.2 | XP_005255309.1 | 7 - 8 | 1452 | 94 | |
XM_017023135.1 | XP_016878624.1 | 5 - 6 | 1209 | 94 | |
GenBank mRNA | AB105376.1 | - | 7 - 8 | 1081 | 94 |
AJ245820.1 | - | 7 - 8 | 1081 | 94 | |
AJ245821.1 | - | 7 - 8 | 1081 | 94 | |
AJ245822.1 | - | 7 - 8 | 1081 | 94 | |
AK027407.1 | - | 5 - 6 | 722 | 94 | |
AK027676.1 | - | 4 - 5 | 692 | 94 | |
AK074998.1 | - | 3 - 4 | 410 | 94 | |
AK299103.1 | - | 7 - 8 | 1449 | 94 | |
AY358404.1 | - | 7 - 8 | 1441 | 94 | |
BC000567.2 | - | 5 - 6 | 910 | 94 | |
KJ534938.1 | - | 7 - 8 | 1207 | 94 | |
KJ534939.1 | - | 5 - 6 | 871 | 94 | |
KJ534940.1 | - | 7 - 8 | 1207 | 94 | |
KJ534941.1 | - | 5 - 6 | 865 | 94 | |
KJ534988.1 | - | 7 - 8 | 1213 | 94 | |
KJ535068.1 | - | 7 - 8 | 1118 | 94 | |
KJ535092.1 | - | 7 - 8 | 1206 | 94 |