Assay Design



Gene Transcripts

Gene Symbol
INPP5F
Entrez Gene ID
22876
Gene Name
inositol polyphosphate-5-phosphatase F
Gene Aliases
MSTP007, MSTPO47, SAC2, hSAC2
Chromosome Location
Chr.10: 119704395 - 119829151 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001243194.1 NP_001230123.1 1 - 2 279 142
GenBank mRNA AK091448.1 - 1 - 2 819 142
AK123633.1 - 1 - 2 262 142
CD642547.1 - 1 - 2 260 142
CN362016.1 - 1 - 2 281 142
DN992317.1 - 1 - 2 203 142


Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA CN362025.1 - 1 - 2 255 Chr. 10 - 119819284 - 119823183 [+] on Build GRCh38 142
GenBankmRNA CN362024.1 - 1 - 2 239 Chr. 10 - 119819297 - 119823900 [+] on Build GRCh38 142
GenBankmRNA CD643666.1 - 1 - 2 210 Chr. 10 - 119819337 - 119826747 [+] on Build GRCh38 142
GenBankmRNA DA879187.1 - 1 - 2 209 Chr. 10 - 119819327 - 119823896 [+] on Build GRCh38 142
GenBankmRNA BU580074.1 - 1 - 2 209 Chr. 10 - 119819364 - 119823138 [+] on Build GRCh38 142
GenBankmRNA DA574365.1 - 1 - 2 264 Chr. 10 - 119819272 - 119823202 [+] on Build GRCh38 142
GenBankmRNA CN362017.1 - 1 - 2 199 Chr. 10 - 119819337 - 119823183 [+] on Build GRCh38 142
GenBankmRNA CN362019.1 - 1 - 2 235 Chr. 10 - 119819301 - 119823135 [+] on Build GRCh38 142
GenBankmRNA CN362021.1 - 1 - 2 156 Chr. 10 - 119819380 - 119823137 [+] on Build GRCh38 142
GenBankmRNA BQ722474.1 - 1 - 2 186 Chr. 10 - 119819350 - 119826958 [+] on Build GRCh38 142
GenBankmRNA CN362041.1 - 1 - 2 235 Chr. 10 - 119819301 - 119823139 [+] on Build GRCh38 142
GenBankmRNA CN362042.1 - 1 - 2 111 Chr. 10 - 119819425 - 119823170 [+] on Build GRCh38 142
GenBankmRNA CD642791.1 - 1 - 2 210 Chr. 10 - 119819337 - 119826819 [+] on Build GRCh38 142
GenBankmRNA DA486507.1 - 1 - 2 122 Chr. 10 - 119819414 - 119826711 [+] on Build GRCh38 142
GenBankmRNA CD656252.1 - 1 - 2 191 Chr. 10 - 119819355 - 119826857 [+] on Build GRCh38 142
GenBankmRNA M78501.1 - 1 - 2 175 Chr. 10 - 119819361 - 119822480 [+] on Build GRCh38 142
GenBankmRNA BQ719336.1 - 176 Chr. - - [-] on Build 142
GenBankmRNA DA500367.1 - 1 - 2 208 Chr. 10 - 119819328 - 119823904 [+] on Build GRCh38 142
GenBankmRNA AL133757.1 - 1 - 2 197 Chr. 10 - 119819339 - 119820878 [+] on Build GRCh38 142


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream BCL2 associated athanogene 3 BAG3 BAG-3, BIS, CAIR-1, MFM6 -141.4kb Sense Chr. - 119651347 - 119677819 on Build GRCh38
Downstream minichromosome maintenance complex binding protein MCMBP C10orf119, MCM-BP 8.5kb Anti-sense Chr. - 119829404 - 119873628 on Build GRCh38

View Full Product Details