Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_003584.3 | 2 - 3 | 423 | 78 | |
NR_034010.1 | 3 - 4 | 272 | 78 | ||
NR_034011.1 | 2 - 3 | 78 | 78 | ||
GenBank mRNA | BC017993.1 | - | 2 - 3 | 429 | 78 |
BM845860.1 | - | 3 - 4 | 277 | 78 | |
CD520851.1 | - | 2 - 3 | 85 | 78 | |
CF124302.1 | - | 2 - 3 | 466 | 78 |