Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_003921.1 | 1 - 2 | 227 | 152 | |
GenBank mRNA | AY008407.1 | - | 1 - 2 | 227 | 152 |
AY008408.1 | - | 1 - 2 | 227 | 152 | |
AY008409.1 | - | 1 - 2 | 227 | 152 | |
AY008410.1 | - | 1 - 2 | 227 | 152 | |
EU921426.1 | - | 1 - 2 | 97 | 152 | |
EU921427.1 | - | 1 - 2 | 97 | 152 | |
EU921428.1 | - | 1 - 2 | 152 | ||
EU921429.1 | - | 1 - 2 | 97 | 152 | |
EU921430.1 | - | 1 - 2 | 97 | 152 |
Gene Name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromosome | |
---|---|---|---|---|---|---|
Upstream | glucuronidase, beta pseudogene 1 | GUSBP1 | -553.0kb | Sense | Chr. - 21459480 - 21589372 on Build GRCh38 | |
Downstream | PR domain 9 | PRDM9 | MEISETZ, MSBP3, PFM6, ZNF899 | 1.4mb | Sense | Chr. - 23507609 - 23528597 on Build GRCh38 |