Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_022014.1 | 5 - 6 | 673 | 79 | |
GenBank mRNA | AY271964.1 | - | 5 - 6 | 659 | 79 |
AY271967.1 | - | 3 - 4 | 413 | 79 |
Gene Name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromosome | |
---|---|---|---|---|---|---|
Upstream | solute carrier family 30 member 4 | SLC30A4 | ZNT4, znT-4 | -23.4kb | Anti-sense | Chr. - 45479613 - 45523685 on Build GRCh38 |
Downstream | biogenesis of lysosomal organelles complex 1 subunit 6 | BLOC1S6 | BLOS6, HPS9, PA, PALLID, PLDN | 30.5kb | Sense | Chr. - 45587123 - 45609716 on Build GRCh38 |