Assay Design
Gene Transcripts
- Gene Symbol
- SNORA38
- Entrez Gene ID
-
677820
- Gene Name
- small nucleolar RNA, H/ACA box 38
- Gene Aliases
-
ACA38
- Chromosome Location
-
Chr.6: 31623079 - 31623210 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_002971.1 |
|
1 - 1 |
95 |
111 |
GenBank mRNA |
AJ609474.1 |
- |
1 - 1 |
95 |
111 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
proline rich coiled-coil 2A |
PRRC2A |
BAT2, D6S51, D6S51E, G2 |
|
Sense |
Chr. - 31620673 - 31637780 on Build GRCh38 |
Downstream |
microRNA 6832 |
MIR6832 |
hsa-mir-6832 |
10.6kb
|
Sense |
Chr. - 31633787 - 31633858 on Build GRCh38 |
View Full Product Details