Assay Design
Gene Transcripts
- Gene Symbol
- SNORA11C
- Entrez Gene ID
-
100124540
- Gene Name
- small nucleolar RNA, H/ACA box 11C
- Gene Aliases
-
-
- Chromosome Location
-
Chr.X: 47388650 - 47388776 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_003710.1 |
|
1 - 1 |
87 |
82 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
ubiquitin specific peptidase 11 |
USP11 |
UHX1 |
-140.3kb
|
Sense |
Chr. - 47232915 - 47248328 on Build GRCh38 |
Downstream |
zinc finger protein 41 |
ZNF41 |
MRX89 |
56.0kb
|
Anti-sense |
Chr. - 47444691 - 47483671 on Build GRCh38 |
View Full Product Details