Product Details
- Assay Reference Genome
Location
Chr.6:21688343 on build GRCh38
- Cytoband
- 6p22.3
- Species
- Homo sapiens
- Variation Type
- Copy Number
Additional Information
For this assay, SNP(s) [rs78278852] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Target Gene Details
- Entrez Gene ID
- 401237
- Gene Symbol
- CASC15
- Gene Name
- cancer susceptibility candidate 15 (non-protein coding)
- Gene Aliases
- LINC00340
- Location
- Chr.6:21666444-22194400 on build GRCh38
- Assay Gene Location
- Within Intron 2
View Full Product Details