Product Details

Assay Reference Genome
Location

Chr.7:44146393 on build GRCh38
Cytoband
7p13
Species
Homo sapiens
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
2645
Gene Symbol
GCK
Gene Name
glucokinase
Gene Aliases
FGQTL3, GK, GLK, HHF3, HK4, HKIV, HXKP, LGLK, MODY2
Location
Chr.7:44143213-44189439 on build GRCh38
Assay Gene Location
Within Intron 10
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
GCK NM_000162.3 NP_000153.1
NM_033507.1 NP_277042.1
NM_033508.1 NP_277043.1
XM_017011966.1 XP_016867455.1
AK122876.1
BC001890.1 AAH01890.1
BT007062.1 AAP35711.1
EU008605.1
M69051.1 AAB59563.1
M88011.1 AAA51824.1
M90299.1 AAA52562.1

Target Copy Number Variation Details

DGV Version
Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv830983 Chr7:44068842 - 44201624 on Build GRCh38 Loss POLM MYL7 GCK MIR4649 YKT6 MIR6838 POLD2 AEBP1
nsv8082 Chr7:43956156 - 44207229 on Build GRCh38 Gain LINC00957 RASA4CP PGAM2 YKT6 POLR2J4 POLM MYL7 SPDYE1 GCK MIR6837 MIR4649 DBNL MIR6838 POLD2 AEBP1
nsv428165 Chr7:43855828 - 44172960 on Build GRCh38 Gain LINC00957 LOC107986791 RASA4CP URGCP PGAM2 POLR2J4 UBE2D4 POLM MYL7 SPDYE1 GCK MIR6837 MIR4649 URGCP-MRPS24 DBNL MIR6838 MRPS24 POLD2 AEBP1

View Full Product Details