Product Details

Assay Reference Genome
Location

Chr.7:44150267 on build GRCh38
Cytoband
7p13
Species
Homo sapiens
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
2645
Gene Symbol
GCK
Gene Name
glucokinase
Gene Aliases
FGQTL3, GK, GLK, HHF3, HK4, HKIV, HXKP, LGLK, MODY2
Location
Chr.7:44143213-44189439 on build GRCh38
Assay Gene Location
Within Intron 6
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
GCK NM_000162.3 NP_000153.1
NM_033507.1 NP_277042.1
NM_033508.1 NP_277043.1
XM_017011966.1 XP_016867455.1
AK122876.1
BC001890.1 AAH01890.1
BT007062.1 AAP35711.1
EU008605.1
M69051.1 AAB59563.1
M88011.1 AAA51824.1
M90299.1 AAA52562.1

Target Copy Number Variation Details

DGV Version
Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv830983 Chr7:44068842 - 44201624 on Build GRCh38 Loss MYL7 YKT6 AEBP1 MIR4649 GCK MIR6838 POLD2 POLM
nsv8082 Chr7:43956156 - 44207229 on Build GRCh38 Gain MYL7 PGAM2 AEBP1 GCK MIR6838 POLD2 POLM LINC00957 YKT6 RASA4CP MIR4649 MIR6837 SPDYE1 POLR2J4 DBNL
nsv428165 Chr7:43855828 - 44172960 on Build GRCh38 Gain UBE2D4 MYL7 PGAM2 AEBP1 GCK MIR6838 POLD2 POLM LOC107986791 LINC00957 URGCP-MRPS24 RASA4CP MIR4649 URGCP MIR6837 SPDYE1 POLR2J4 MRPS24 DBNL
nsv509195 Chr7:44147118 - 44261782 on Build GRCh38 Insertion YKT6 GCK CAMK2B

View Full Product Details