Assay Design



Gene Transcripts

Gene Symbol
STEAP1B
Entrez Gene ID
256227
Gene Name
STEAP family member 1B
Gene Aliases
-
Chromosome Location
Chr.7: 22419444 - 22500282 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001164460.1 NP_001157932.1 4 - 5 959 86
GenBank mRNA AK308991.1 - 4 - 5 856 86

View Full Product Details