Assay Design
Gene Transcripts
- Gene Symbol
- SDHAP3
- Entrez Gene ID
-
728609
- Gene Name
- succinate dehydrogenase complex flavoprotein subunit A pseudogene 3
- Gene Aliases
-
SDHACL, SDHAL
- Chromosome Location
-
Chr.5: 1571957 - 1594531 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_003263.2 |
|
4 - 5 |
570 |
61 |
View Full Product Details