Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_024035.2 | 9 - 10 | 1300 | 85 | |
NR_135095.1 | 9 - 10 | 1303 | 85 | ||
GenBank mRNA | BC140764.1 | - | 9 - 10 | 85 | |
BC144474.1 | - | 9 - 10 | 85 |
Gene Name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromosome | |
---|---|---|---|---|---|---|
Upstream | zinc finger protein 354A | ZNF354A | EZNF, HEL104, HKL1, KID-1, KID1, TCF17 | -36.4kb | Sense | Chr. - 178711521 - 178730702 on Build GRCh38 |
Downstream | zinc finger protein 354B | ZNF354B | KID2 | 91.3kb | Anti-sense | Chr. - 178859813 - 178901135 on Build GRCh38 |