Assay Design
Gene Transcripts
- Gene Symbol
- LOC400620
- Entrez Gene ID
-
400620
- Gene Name
- uncharacterized LOC400620
- Gene Aliases
-
-
- Chromosome Location
-
Chr.17: 73786822 - 73800795 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_040020.1 |
|
1 - 2 |
999 |
98 |
GenBank mRNA |
BC035399.1 |
- |
1 - 2 |
999 |
98 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
uncharacterized LOC100134391 |
LOC100134391 |
|
-30.0kb
|
Sense |
Chr. - 73737854 - 73757723 on Build GRCh38 |
Downstream |
ribosomal protein L38 |
RPL38 |
L38 |
413.5kb
|
Sense |
Chr. - 74203656 - 74209880 on Build GRCh38 |
View Full Product Details