Assay Design
Gene Transcripts
- Gene Symbol
- PTEN
- Entrez Gene ID
-
5728
- Gene Name
- phosphatase and tensin homolog
- Gene Aliases
-
10q23del, BZS, CWS1, DEC, GLM2, MHAM, MMAC1, PTEN1, TEP1
- Chromosome Location
-
Chr.10: 87863438 - 87971930 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK124525.1 |
- |
1 - 1 |
2455 |
77 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
killin, p53-regulated DNA replication inhibitor |
KLLN |
CWS4, KILLIN |
-24.1kb
|
Anti-sense |
Chr. - 87859161 - 87863437 on Build GRCh38 |
Downstream |
renalase, FAD dependent amine oxidase |
RNLS |
C10orf59, RENALASE |
288.7kb
|
Anti-sense |
Chr. - 88176458 - 88583860 on Build GRCh38 |
View Full Product Details