Assay Design
Gene Transcripts
- Gene Symbol
- C9orf3
- Entrez Gene ID
-
84909
- Gene Name
- chromosome 9 open reading frame 3
- Gene Aliases
-
AOPEP, AP-O, APO, C90RF3, ONPEP
- Chromosome Location
-
Chr.9: 94726604 - 95148264 on Build GRCh38
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
CB155975.1 |
- |
1 |
105 |
|
Chr. 9 - 95085787 - 95086313 [+] on Build GRCh38 |
97 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
microRNA 6081 |
MIR6081 |
hsa-mir-6081 |
-17.8kb
|
Sense |
Chr. - 95065350 - 95065446 on Build GRCh38 |
Downstream |
Fanconi anemia complementation group C |
FANCC |
FA3, FAC, FACC |
11.9kb
|
Anti-sense |
Chr. - 95099054 - 95317730 on Build GRCh38 |
View Full Product Details