Assay Design
Gene Transcripts
- Gene Symbol
- AGBL4
- Entrez Gene ID
-
84871
- Gene Name
- ATP/GTP binding protein like 4
- Gene Aliases
-
CCP6
- Chromosome Location
-
Chr.1: 48520157 - 50024011 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AF075049.1 |
- |
1 - 1 |
50 |
111 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
H38784.1 |
- |
1 |
50 |
|
Chr. 1 - 49690436 - 49690829 [-] on Build GRCh38 |
111 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
AGBL4 intronic transcript 1 |
AGBL4-IT1 |
|
-218.3kb
|
Sense |
Chr. - 49374201 - 49472085 on Build GRCh38 |
Downstream |
ELAV like neuron-specific RNA binding protein 4 |
ELAVL4 |
HUD, PNEM |
357.2kb
|
Anti-sense |
Chr. - 50048014 - 50203786 on Build GRCh38 |
View Full Product Details