Assay Design
Gene Transcripts
- Gene Symbol
- LINC00667
- Entrez Gene ID
-
339290
- Gene Name
- long intergenic non-protein coding RNA 667
- Gene Aliases
-
-
- Chromosome Location
-
Chr.18: 5238100 - 5246508 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BC119664.1 |
- |
1 - 2 |
638 |
104 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
A kinase (PRKA) anchor inhibitor 1 |
AKAIN1 |
C18orf42 |
-34.7kb
|
Anti-sense |
Chr. - 5143673 - 5199997 on Build GRCh38 |
Downstream |
long intergenic non-protein coding RNA 526 |
LINC00526 |
C18orf18, HsT959 |
824.0
|
Anti-sense |
Chr. - 5236724 - 5238029 on Build GRCh38 |
View Full Product Details