Assay Design



Gene Transcripts

Gene Symbol
NDUFC1
Entrez Gene ID
4717
Gene Name
NADH:ubiquinone oxidoreductase subunit C1
Gene Aliases
KFYI
Chromosome Location
Chr.4: 139289917 - 139302551 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001184986.1 NP_001171915.1 1 - 1 392 72
NM_001184991.1 NP_001171920.1 1 - 1 392 72
GenBank mRNA AK023115.1 - 1 - 1 1653 72
CX163301.1 - 1 - 1 392 72


Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BM771098.1 - 1 355 Chr. 4 - 139300602 - 139301173 [-] on Build GRCh38 72
GenBankmRNA CN360331.1 - 1 396 Chr. 4 - 139300602 - 139301213 [-] on Build GRCh38 72
GenBankmRNA BM785365.1 - 1 355 Chr. 4 - 139300610 - 139301173 [-] on Build GRCh38 72
GenBankmRNA BU685870.1 - 1 Chr. 4 - 139300419 - 139300899 [+] on Build GRCh38 72
GenBankmRNA AU152690.2 - 1 Chr. 4 - 139300421 - 139300960 [+] on Build GRCh38 72
GenBankmRNA BG676936.1 - 1 510 Chr. 4 - 139300661 - 139301313 [-] on Build GRCh38 72
GenBankmRNA BM784018.1 - 1 355 Chr. 4 - 139295863 - 139301173 [-] on Build GRCh38 72
GenBankmRNA CN360329.1 - 1 384 Chr. 4 - 139295926 - 139301202 [-] on Build GRCh38 72
GenBankmRNA BI822563.1 - 1 237 Chr. 4 - 139295049 - 139301054 [-] on Build GRCh38 72
GenBankmRNA BX477751.1 - 1 486 Chr. 4 - 139300702 - 139301303 [-] on Build GRCh38 72
GenBankmRNA BM747103.1 - 1 355 Chr. 4 - 139300648 - 139301173 [-] on Build GRCh38 72
GenBankmRNA DA895687.1 - 1 518 Chr. 4 - 139300785 - 139301336 [-] on Build GRCh38 72
GenBankmRNA BM745489.1 - 1 355 Chr. 4 - 139295942 - 139301173 [-] on Build GRCh38 72
GenBankmRNA BM756332.1 - 1 355 Chr. 4 - 139300602 - 139301173 [-] on Build GRCh38 72
GenBankmRNA BU600840.1 - 1 204 Chr. 4 - 139295049 - 139301020 [-] on Build GRCh38 72
GenBankmRNA BM768085.1 - 1 355 Chr. 4 - 139300602 - 139301173 [-] on Build GRCh38 72
GenBankmRNA BI488544.1 - 1 578 Chr. 4 - 139300621 - 139301395 [-] on Build GRCh38 72


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream mitochondria localized glutamic acid rich protein MGARP C4orf49, CESP-1, HUMMR, OSAP -20.1kb Sense Chr. - 139266163 - 139280338 on Build GRCh38
Downstream uncharacterized LOC105377450 LOC105377450 91.1kb Anti-sense Chr. - 139393591 - 139420276 on Build GRCh38

View Full Product Details