Assay Design
Gene Transcripts
- Gene Symbol
- HFE
- Entrez Gene ID
-
3077
- Gene Name
- hemochromatosis
- Gene Aliases
-
HFE1, HH, HLA-H, MVCD7, TFQTL2
- Chromosome Location
-
Chr.6: 26087281 - 26096216 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AJ404378.1 |
- |
2 - 2 |
1548 |
114 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
AI760080.1 |
- |
1 |
|
|
Chr. 6 - 26097476 - 26097859 [-] on Build GRCh38 |
114 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Downstream |
histone cluster 1, H4c |
HIST1H4C |
H4/g, H4FG, dJ221C16.1 |
5.6kb
|
Sense |
Chr. - 26103948 - 26104337 on Build GRCh38 |
View Full Product Details