Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
GenBank mRNA | AY328030.1 | - | 3 - 3 | 352 | 88 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
---|---|---|---|---|---|---|---|
GenBankmRNA | BG201709.1 | - | 79 | Chr. - - [-] on Build | 88 |
Gene Name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromosome | |
---|---|---|---|---|---|---|
Upstream | catenin delta 2 | CTNND2 | GT24, NPRAP | -750.6kb | Anti-sense | Chr. - 10971840 - 11904067 on Build GRCh38 |
Downstream | dynein axonemal heavy chain 5 | DNAH5 | CILD3, DNAHC5, HL1, KTGNR, PCD | 1.0mb | Anti-sense | Chr. - 13690328 - 14011829 on Build GRCh38 |