Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
GenBank mRNA | BC029446.1 | - | 1 - 1 | 554 | 99 |
Gene Name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromosome | |
---|---|---|---|---|---|---|
Upstream | proprotein convertase subtilisin/kexin type 1 inhibitor | PCSK1N | BigLEN, PEN, PROSAAS, SAAS, SCG8, SgVIII | -61.5kb | Sense | Chr. - 48831092 - 48835638 on Build GRCh38 |
Downstream | polyglutamine binding protein 1 | PQBP1 | MRX2, MRX55, MRXS3, MRXS8, NPW38, RENS1, SHS | 131.0 | Anti-sense | Chr. - 48897862 - 48903145 on Build GRCh38 |