Assay Design
Gene Transcripts
- Gene Symbol
- PRDM5
- Entrez Gene ID
-
11107
- Gene Name
- PR domain 5
- Gene Aliases
-
BCS2, PFM2
- Chromosome Location
-
Chr.4: 120684907 - 120922866 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BC047115.1 |
- |
1 - 1 |
551 |
95 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
BQ045043.1 |
- |
1 |
|
|
Chr. 4 - 120830569 - 120831160 [+] on Build GRCh38 |
95 |
GenBankmRNA |
BQ045042.1 |
- |
1 |
|
|
Chr. 4 - 120830569 - 120831319 [+] on Build GRCh38 |
95 |
GenBankmRNA |
BI668592.1 |
- |
1 |
556 |
|
Chr. 4 - 120830569 - 120831254 [-] on Build GRCh38 |
95 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
MAD2 mitotic arrest deficient-like 1 (yeast) |
MAD2L1 |
HSMAD2, MAD2 |
-763.7kb
|
Sense |
Chr. - 120059422 - 120066858 on Build GRCh38 |
Downstream |
neuron derived neurotrophic factor |
NDNF |
C4orf31, NORD |
204.4kb
|
Sense |
Chr. - 121035627 - 121072518 on Build GRCh38 |
View Full Product Details