Assay Design
Gene Transcripts
- Gene Symbol
- BPESC1
- Entrez Gene ID
-
60467
- Gene Name
- blepharophimosis, epicanthus inversus and ptosis, candidate 1 (non-protein coding)
- Gene Aliases
-
NCRNA00187
- Chromosome Location
-
Chr.3: 139104185 - 139125167 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_026783.3 |
|
2 - 3 |
2328 |
114 |
GenBank mRNA |
AF196865.1 |
- |
2 - 3 |
2327 |
114 |
View Full Product Details