Assay Design
Gene Transcripts
- Gene Symbol
- TUBBP5
- Entrez Gene ID
-
643224
- Gene Name
- tubulin beta pseudogene 5
- Gene Aliases
-
-
- Chromosome Location
-
Chr.9: 138150113 - 138177433 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_027156.1 |
|
1 - 2 |
189 |
107 |
GenBank mRNA |
AK308459.1 |
- |
1 - 2 |
189 |
107 |
View Full Product Details