Assay Design
Gene Transcripts
- Gene Symbol
- CLCA3P
- Entrez Gene ID
-
9629
- Gene Name
- chloride channel accessory 3, pseudogene
- Gene Aliases
-
CLCA3
- Chromosome Location
-
Chr.1: 86634276 - 86655376 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_024604.1 |
|
4 - 5 |
568 |
99 |
GenBank mRNA |
AF043976.1 |
- |
4 - 5 |
568 |
99 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
chloride channel accessory 4 |
CLCA4 |
CaCC, CaCC2 |
-56.0kb
|
Sense |
Chr. - 86547076 - 86580758 on Build GRCh38 |
Downstream |
uncharacterized LOC105378828 |
LOC105378828 |
|
64.5kb
|
Anti-sense |
Chr. - 86571181 - 86704493 on Build GRCh38 |
View Full Product Details