Product Details

Assay Reference Genome
Location

Chr.9:35657806 on build GRCh38
Cytoband
9p13.3
Species
Homo sapiens
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
6023
Gene Symbol
RMRP
Gene Name
RNA component of mitochondrial RNA processing endoribonuclease
Gene Aliases
CHH, NME1, RMRPR, RRP2
Location
Chr.9:35657751-35658018 on build GRCh38
Assay Gene Location
Within Exon 1
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
RMRP NR_003051.3 1 213
AA593275.1 1 188
DW422801.1 1 229
DW423815.1 1 229

View Full Product Details