Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_183372.5 | NP_899228.4 | 3 - 4 | 837 | 102 |
NR_046188.3 | 3 - 4 | 837 | 102 | ||
GenBank mRNA | AF419617.1 | - | 1 - 2 | 87 | 102 |
AL832622.2 | - | 3 - 4 | 970 | 102 | |
BC136292.1 | - | 3 - 4 | 837 | 102 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001278141.1 | NP_001265070.1 | 4 - 5 | 887 | 102 |
GenBank mRNA | AB033071.1 | - | 3 - 4 | 440 | 102 |