Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_011516215.2 | XP_011514517.1 | 4 - 5 | 639 | 96 |
XM_017012208.1 | XP_016867697.1 | 4 - 5 | 639 | 96 | |
XM_017012209.1 | XP_016867698.1 | 4 - 5 | 639 | 96 | |
XM_017012210.1 | XP_016867699.1 | 4 - 5 | 715 | 96 | |
XM_017012211.1 | XP_016867700.1 | 4 - 5 | 639 | 96 | |
XM_017012212.1 | XP_016867701.1 | 4 - 5 | 429 | 96 | |
XM_017012213.1 | XP_016867702.1 | 4 - 5 | 767 | 96 | |
XM_017012214.1 | XP_016867703.1 | 4 - 5 | 343 | 96 | |
XM_017012215.1 | XP_016867704.1 | 4 - 5 | 639 | 96 | |
GenBank mRNA | EU126604.1 | - | 3 - 4 | 283 | 96 |