Assay Design



Gene Transcripts

Gene Symbol
KPNA7
Entrez Gene ID
402569
Gene Name
karyopherin subunit alpha 7
Gene Aliases
IPOA8
Chromosome Location
Chr.7: 99145467 - 99219417 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_011516215.2 XP_011514517.1 10 - 11 66
GenBank mRNA AW193472.1 - 1 - 2 226 66
EU126604.1 - 9 - 10 66

View Full Product Details