Assay Design



Gene Transcripts

Gene Symbol
FCRL1
Entrez Gene ID
115350
Gene Name
Fc receptor like 1
Gene Aliases
CD307a, FCRH1, IFGP1, IRTA5
Chromosome Location
Chr.1: 157792249 - 157820150 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001159397.1 NP_001152869.1 6 - 7 1027 133
XM_005244866.2 XP_005244923.1 7 - 8 1115 133
XM_011509128.1 XP_011507430.1 7 - 8 1115 133
XM_011509129.1 XP_011507431.1 7 - 8 1115 133
XM_011509130.1 XP_011507432.1 7 - 8 1115 133
GenBank mRNA AK096690.1 - 6 - 7 966 133
AL833674.1 - 3 - 4 198 133

View Full Product Details