Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_152864.3 | NP_690603.3 | 4 - 5 | 558 | 70 |
XM_005260192.1 | XP_005260249.1 | 4 - 5 | 520 | 70 | |
XM_005260194.3 | XP_005260251.1 | 4 - 5 | 979 | 70 | |
XM_011528527.1 | XP_011526829.1 | 4 - 5 | 520 | 70 | |
XM_011528528.2 | XP_011526830.1 | 4 - 5 | 982 | 70 | |
XM_011528529.2 | XP_011526831.1 | 4 - 5 | 968 | 70 | |
XM_017027636.1 | XP_016883125.1 | 4 - 5 | 490 | 70 | |
XM_017027637.1 | XP_016883126.1 | 4 - 5 | 972 | 70 | |
XM_017027638.1 | XP_016883127.1 | 4 - 5 | 978 | 70 | |
XM_017027639.1 | XP_016883128.1 | 4 - 5 | 964 | 70 | |
GenBank mRNA | AK126246.1 | - | 4 - 5 | 481 | 70 |
AK225765.1 | - | 4 - 5 | 512 | 70 | |
BC041812.1 | - | 4 - 5 | 556 | 70 |