Assay Design
Gene Transcripts
A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
AF090935.1 |
- |
2 - 3 |
1126 |
|
Chr. 20 - 24962944 - 24992642 [+] on Build GRCh38 |
98 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
adipocyte plasma membrane associated protein |
APMAP |
BSCv, C20orf3 |
|
Anti-sense |
Chr. - 24962925 - 24992974 on Build GRCh38 |
Downstream |
acyl-CoA synthetase short-chain family member 1 |
ACSS1 |
ACAS2L, ACECS1, AceCS2L |
37.1kb
|
Anti-sense |
Chr. - 25006230 - 25058182 on Build GRCh38 |
View Full Product Details