Assay Design
Gene Transcripts
- Gene Symbol
- TSNAX-DISC1
- Entrez Gene ID
-
100303453
- Gene Name
- TSNAX-DISC1 readthrough (NMD candidate)
- Gene Aliases
-
-
- Chromosome Location
-
Chr.1: 231528653 - 232041272 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AF230316.1 |
- |
1 - 1 |
152 |
101 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
long intergenic non-protein coding RNA 582 |
LINC00582 |
|
-5.7kb
|
Anti-sense |
Chr. - 231591292 - 231612090 on Build GRCh38 |
Downstream |
disrupted in schizophrenia 1 |
DISC1 |
C1orf136, SCZD9 |
8.8kb
|
Sense |
Chr. - 231626815 - 232041272 on Build GRCh38 |
View Full Product Details