Assay Design
Gene Transcripts
A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
AK000114.1 |
- |
1 |
745 |
|
Chr. 16 - 8634771 - 8636543 [+] on Build GRCh38 |
79 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
methyltransferase like 22 |
METTL22 |
C16orf68 |
|
Sense |
Chr. - 8621640 - 8673565 on Build GRCh38 |
Downstream |
4-aminobutyrate aminotransferase |
ABAT |
GABA-AT, GABAT, NPD009 |
38.0kb
|
Sense |
Chr. - 8674587 - 8784575 on Build GRCh38 |
View Full Product Details