Assay Design
Gene Transcripts
A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
AL137323.1 |
- |
1 |
4350 |
|
Chr. 1 - 54138373 - 54142980 [+] on Build GRCh38 |
67 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
CUB domain containing protein 2 |
CDCP2 |
|
|
Anti-sense |
Chr. - 54138995 - 54153006 on Build GRCh38 |
Downstream |
cytochrome b5 reductase like |
CYB5RL |
|
29.4kb
|
Anti-sense |
Chr. - 54172336 - 54200073 on Build GRCh38 |
View Full Product Details