Assay Design



Gene Transcripts

Gene Symbol
PEX11A
Entrez Gene ID
8800
Gene Name
peroxisomal biogenesis factor 11 alpha
Gene Aliases
PEX11-ALPHA, PMP28, hsPEX11p
Chromosome Location
Chr.15: 89681531 - 89690784 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001271572.1 NP_001258501.1 3 - 3 2343 66
NM_001271573.1 NP_001258502.1 2 - 2 2320 66
NM_003847.2 NP_003838.1 3 - 3 2436 66
GenBank mRNA AI494060.1 - 1 - 1 66
AL109728.1 - 1 - 1 548 66
AL360141.1 - 1 - 1 1550 66


Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BX412834.1 - 1 Chr. 15 - 89681561 - 89682385 [+] on Build GRCh38 66
GenBankmRNA CA308833.1 - 1 Chr. 15 - 89681535 - 89682233 [+] on Build GRCh38 66
GenBankmRNA AA703022.1 - 1 Chr. 15 - 89681541 - 89682034 [+] on Build GRCh38 66
GenBankmRNA BU617530.1 - 1 Chr. 15 - 89681535 - 89682266 [+] on Build GRCh38 66
GenBankmRNA BQ437764.1 - 1 326 Chr. 15 - 89681535 - 89682162 [-] on Build GRCh38 66
GenBankmRNA BQ613754.1 - 1 Chr. 15 - 89681555 - 89682049 [+] on Build GRCh38 66
GenBankmRNA BQ007145.1 - 1 Chr. 15 - 89681531 - 89682252 [+] on Build GRCh38 66
GenBankmRNA AI254741.1 - 1 Chr. 15 - 89681760 - 89682124 [+] on Build GRCh38 66
GenBankmRNA AI866665.1 - 1 Chr. 15 - 89681539 - 89682080 [+] on Build GRCh38 66
GenBankmRNA AI016995.1 - 1 Chr. 15 - 89681536 - 89681882 [+] on Build GRCh38 66
GenBankmRNA AI676167.1 - 1 Chr. 15 - 89681535 - 89681976 [+] on Build GRCh38 66
GenBankmRNA AI245303.1 - 1 Chr. 15 - 89681539 - 89681966 [+] on Build GRCh38 66
GenBankmRNA BX434749.1 - 1 Chr. 15 - 89681559 - 89682389 [+] on Build GRCh38 66
GenBankmRNA AA889745.1 - 1 Chr. 15 - 89681538 - 89681953 [+] on Build GRCh38 66


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream perilipin 1 PLIN1 FPLD4, PERI, PLIN -2.1kb Sense Chr. - 89664365 - 89679417 on Build GRCh38
Downstream WD repeat domain 93 WDR93 6.7kb Anti-sense Chr. - 89690074 - 89744390 on Build GRCh38

View Full Product Details