Assay Design



Gene Transcripts

A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.



Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA N45115.1 - 1 57 Chr. 4 - 169723518 - 169724123 [+] on Build GRCh38 86
GenBankmRNA BM676149.1 - 1 Chr. 4 - 169723011 - 169723673 [-] on Build GRCh38 86
GenBankmRNA BX095552.1 - 1 Chr. 4 - 169723513 - 169724245 [-] on Build GRCh38 86
GenBankmRNA G36553.1 - 1 57 Chr. 4 - 169723518 - 169724012 [+] on Build GRCh38 86


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream chloride voltage-gated channel 3 CLCN3 CLC3, ClC-3 -331.0 Sense Chr. - 169620521 - 169723187 on Build GRCh38
Downstream histone PARylation factor 1 HPF1 C4orf27 5.5kb Anti-sense Chr. - 169729468 - 169757954 on Build GRCh38

View Full Product Details