Assay Design



Gene Transcripts

A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.



Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA AJ011601.1 - 1 147 Chr. 11 - 27695459 - 27695686 [+] on Build GRCh38 91


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream long intergenic non-protein coding RNA 678 LINC00678 -60.8kb Anti-sense Chr. - 27617626 - 27634627 on Build GRCh38
Downstream heat shock protein 90 alpha family class A member 2, pseudogene HSP90AA2P HSP90AA2, HSP90ALPHA, HSPCA, HSPCAL3 192.5kb Anti-sense Chr. - 27888171 - 27891092 on Build GRCh38

View Full Product Details