Assay Design
Gene Transcripts
- Gene Symbol
- INVS
- Entrez Gene ID
-
27130
- Gene Name
- inversin
- Gene Aliases
-
INV, NPH2, NPHP2
- Chromosome Location
-
Chr.9: 100099185 - 100302175 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BC110864.1 |
- |
1 - 2 |
142 |
61 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
BM804824.1 |
- |
1 - 2 |
165 |
|
Chr. 9 - 100099275 - 100304927 [+] on Build GRCh38 |
61 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
endoplasmic reticulum protein 44 |
ERP44 |
PDIA10, TXNDC4 |
-223.0
|
Anti-sense |
Chr. - 99979179 - 100099052 on Build GRCh38 |
Downstream |
testis expressed 10 |
TEX10 |
Ipi1, bA208F1.2 |
200.1kb
|
Anti-sense |
Chr. - 100302075 - 100352977 on Build GRCh38 |
View Full Product Details