Assay Design
Gene Transcripts
A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
BC128395.1 |
- |
1 |
2553 |
|
Chr. 16 - 70713987 - 70716894 [+] on Build GRCh38 |
78 |
GenBankmRNA |
GD259990.1 |
- |
1 |
514 |
|
Chr. 16 - 70716019 - 70716588 [+] on Build GRCh38 |
78 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
metastasis suppressor 1 like |
MTSS1L |
ABBA-1, ABBA1 |
-27.9kb
|
Anti-sense |
Chr. - 70661204 - 70686207 on Build GRCh38 |
Downstream |
VAC14 antisense RNA 1 |
VAC14-AS1 |
|
38.2kb
|
Sense |
Chr. - 70755060 - 70773251 on Build GRCh38 |
View Full Product Details