Assay Design
Gene Transcripts
A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
DQ656066.1 |
- |
2 - 3 |
677 |
|
Chr. 11 - 5249468 - 5645789 [+] on Build GRCh38 |
115 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
olfactory receptor family 51 subfamily B member 2 (gene/pseudogene) |
OR51B2 |
HOR5'Beta3, OR51B1P |
|
Anti-sense |
Chr. - 5323298 - 5324352 on Build GRCh38 |
Downstream |
olfactory receptor family 51 subfamily B member 6 |
OR51B6 |
HOR5'Beta6 |
5.6kb
|
Sense |
Chr. - 5351508 - 5352446 on Build GRCh38 |
View Full Product Details