Assay Design
Gene Transcripts
- Gene Symbol
- SEC22B
- Entrez Gene ID
-
9554
- Gene Name
- SEC22 homolog B, vesicle trafficking protein (gene/pseudogene)
- Gene Aliases
-
ERS-24, SEC22L1
- Chromosome Location
-
Chr.1: 120152520 - 120176534 on Build GRCh38
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
DB216452.1 |
- |
1 |
470 |
|
Chr. 1 - 120158193 - 120158706 [-] on Build GRCh38 |
85 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
notch 2 |
NOTCH2 |
AGS2, HJCYS, hN2 |
-87.6kb
|
Sense |
Chr. - 119911553 - 120073449 on Build GRCh38 |
Downstream |
neuroblastoma breakpoint family member 8 |
NBPF8 |
NBPF8P |
257.2kb
|
Anti-sense |
Chr. - 120436351 - 120467844 on Build GRCh38 |
View Full Product Details