Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_005244843.3 | XP_005244900.1 | 16 - 16 | 7766 | 72 |
XM_011509079.2 | XP_011507381.1 | 15 - 15 | 7700 | 72 | |
XM_011509080.2 | XP_011507382.1 | 14 - 14 | 7534 | 72 | |
XM_017000084.1 | XP_016855573.1 | 16 - 16 | 7787 | 72 | |
GenBank mRNA | AK023363.1 | - | 1 - 1 | 1455 | 72 |
Gene Name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromosome | |
---|---|---|---|---|---|---|
Upstream | SWT1, RNA endoribonuclease homolog | SWT1 | C1orf26, HsSwt1 | -603.0 | Anti-sense | Chr. - 185157023 - 185291781 on Build GRCh38 |