Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
---|---|---|---|---|---|---|---|
GenBankmRNA | BM541932.1 | - | 110 | Chr. - - [-] on Build | 91 |
Gene Name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromosome | |
---|---|---|---|---|---|---|
Upstream | ubiquitin specific peptidase 13 (isopeptidase T-3) | USP13 | ISOT3, IsoT-3 | -5.6kb | Anti-sense | Chr. - 179653145 - 179789401 on Build GRCh38 |
Downstream | PEX5L antisense RNA 2 | PEX5L-AS2 | 100.1kb | Anti-sense | Chr. - 179898194 - 179921899 on Build GRCh38 |