Assay Design



Gene Transcripts

A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.



Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA AF052168.1 - 1 1051 Chr. 1 - 14820008 - 14821530 [+] on Build GRCh38 102


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream kazrin, periplakin interacting protein KAZN KAZ Sense Chr. - 13893387 - 15118048 on Build GRCh38
Downstream TMEM51 antisense RNA 1 TMEM51-AS1 C1orf126 290.3kb Anti-sense Chr. - 15111815 - 15152464 on Build GRCh38

View Full Product Details