Assay Design
Gene Transcripts
- Gene Symbol
- DLGAP1
- Entrez Gene ID
-
9229
- Gene Name
- DLG associated protein 1
- Gene Aliases
-
DAP-1, DAP-1-ALPHA, DAP-1-BETA, DAP1, DLGAP1A, DLGAP1B, GKAP, SAPAP1
- Chromosome Location
-
Chr.18: 3496032 - 4455307 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BC030096.1 |
- |
1 - 1 |
845 |
100 |
BC045785.1 |
- |
1 - 1 |
840 |
100 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
uncharacterized LOC105371967 |
LOC105371967 |
|
-60.8kb
|
Anti-sense |
Chr. - 3653056 - 3656282 on Build GRCh38 |
Downstream |
DLGAP1 antisense RNA 3 |
DLGAP1-AS3 |
|
160.0kb
|
Anti-sense |
Chr. - 3878180 - 3897069 on Build GRCh38 |
View Full Product Details